Assay Details
| Gene Symbol: Mest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252292.1 | NP_001239221.1 | 10 - 11 | 1108 | 87 | |
| NM_001252293.1 | NP_001239222.1 | 9 - 10 | 829 | 87 | ||
| NM_008590.2 | NP_032616.1 | 9 - 10 | 1007 | 87 | ||
| GenBank mRNA | AF482999.1 | - | 10 - 11 | 829 | 87 | |
| AK031718.1 | - | 9 - 10 | 1005 | 87 | ||
| AK032881.1 | - | 9 - 10 | 1006 | 87 | ||
| AK034949.1 | - | 9 - 10 | 1005 | 87 | ||
| AK146457.1 | - | 10 - 11 | 1109 | 87 | ||
| AK168714.1 | - | 9 - 10 | 1005 | 87 | ||
| AK168732.1 | - | 9 - 10 | 1005 | 87 | ||
| AK168743.1 | - | 9 - 10 | 1005 | 87 | ||
| AK168995.1 | - | 9 - 10 | 1005 | 87 | ||
| AK169095.1 | - | 9 - 10 | 1005 | 87 | ||
| AK169266.1 | - | 9 - 10 | 1005 | 87 | ||
| BC006639.1 | - | 9 - 10 | 990 | 3482928 | 87 | |
| D16262.1 | - | 9 - 10 | 994 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mest
Genomic Map