Assay Details
| Gene Symbol: Opn1sw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007538.3 | NP_031564.1 | 2 - 3 | 616 | 117 | |
| GenBank mRNA | AF190670.1 | - | 2 - 3 | 578 | 117 | |
| BC026021.1 | - | 2 - 3 | 583 | 4511808 | 117 | |
| BC050002.1 | - | 2 - 3 | 534 | 117 | ||
| BC058267.1 | - | 2 - 3 | 564 | 6489727 | 117 | |
| U49720.1 | - | 2 - 3 | 518 | 117 | ||
More Information
Set Membership: |
|

Opn1sw
Genomic Map