Assay Details
| Gene Symbol: Impdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047537.1 | - | 10 - 11 | 1290 | 79 | |
| AK089383.1 | - | 10 - 11 | 1254 | 79 | ||
| AK171139.1 | - | 10 - 11 | 1265 | 79 | ||
| AK195203.1 | - | 2 - 3 | 203 | 79 | ||
| AK195247.1 | - | 2 - 3 | 203 | 79 | ||
| AK210877.1 | - | 3 - 4 | 204 | 79 | ||
| BC053416.1 | - | 9 - 10 | 1114 | 6533681 | 79 | |
| U00978.1 | - | 9 - 10 | 1009 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Impdh1
Genomic Map