Assay Details
| Gene Symbol: Impdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047537.1 | - | 5 - 6 | 598 | 63 | |
| AK089383.1 | - | 5 - 6 | 562 | 63 | ||
| AK171139.1 | - | 5 - 6 | 573 | 63 | ||
| AK199145.1 | - | 2 - 3 | 118 | 63 | ||
| BC053416.1 | - | 4 - 5 | 422 | 6533681 | 63 | |
| U00978.1 | - | 4 - 5 | 317 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Impdh1
Genomic Map