Assay Details
| Gene Symbol: Impdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047537.1 | - | 8 - 9 | 1106 | 63 | |
| AK089383.1 | - | 8 - 9 | 1070 | 63 | ||
| AK171139.1 | - | 8 - 9 | 1081 | 63 | ||
| BC053416.1 | - | 7 - 8 | 930 | 6533681 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Impdh1
Genomic Map