Assay Details
| Gene Symbol: Ing3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023626.4 | NP_076115.3 | 1 - 2 | 181 | 70 | |
| GenBank mRNA | AK080787.1 | - | 1 - 2 | 150 | 70 | |
| AK131993.1 | - | 1 - 2 | 144 | 70 | ||
| AY007791.1 | - | 1 - 2 | 150 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK080787.1, AY007791.1, NM_023626.4 transcripts.
Set Membership: |
|

Ing3
Genomic Map