Assay Details
| Gene Symbol: Ing3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023626.4 | NP_076115.3 | 2 - 3 | 252 | 102 | |
| GenBank mRNA | AK030364.1 | - | 1 - 2 | 59 | 102 | |
| AK080787.1 | - | 2 - 3 | 221 | 102 | ||
| AY007791.1 | - | 2 - 3 | 221 | 102 | ||
| BC018342.1 | - | 2 - 3 | 202 | 4010253 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ing3
Genomic Map