Assay Details
| Gene Symbol: Ing3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023626.4 | NP_076115.3 | 3 - 4 | 355 | 77 | |
| GenBank mRNA | AK030364.1 | - | 2 - 3 | 162 | 77 | |
| AK080787.1 | - | 3 - 4 | 324 | 77 | ||
| AK131993.1 | - | 3 - 4 | 355 | 77 | ||
| AK146145.1 | - | 2 - 3 | 172 | 77 | ||
| AY007791.1 | - | 3 - 4 | 324 | 77 | ||
| BC018342.1 | - | 3 - 4 | 305 | 4010253 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ing3
Genomic Map