Assay Details
| Gene Symbol: Rala | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019491.5 | NP_062364.3 | 3 - 4 | 558 | 91 | |
| GenBank mRNA | AF244951.1 | - | 2 - 3 | 321 | 91 | |
| AK013881.1 | - | 3 - 4 | 547 | 91 | ||
| BC031741.1 | - | 3 - 4 | 558 | 4501311 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rala
Genomic Map