Assay Details
| Gene Symbol: Zkscan3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145778.1 | NP_001139250.1 | 2 - 3 | 558 | 58 | |
| NM_023685.4 | NP_076174.3 | 2 - 3 | 555 | 58 | ||
| GenBank mRNA | AK028902.1 | - | 2 - 3 | 545 | 58 | |
| AK031080.1 | - | 2 - 3 | 516 | 58 | ||
| AK031286.1 | - | 2 - 3 | 513 | 58 | ||
| AK050028.1 | - | 2 - 3 | 517 | 58 | ||
| AK220177.1 | - | 2 - 3 | 723 | 58 | ||
| BC007473.1 | - | 2 - 3 | 513 | 3496252 | 58 | |
| BG296820.1 | - | 1 - 2 | 51 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zkscan3
Genomic Map