Assay Details
| Gene Symbol: Gpld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008156.2 | NP_032182.2 | 10 - 11 | 994 | 117 | |
| GenBank mRNA | AK004957.2 | - | 9 - 10 | 698 | 117 | |
| AY081194.1 | - | 9 - 10 | 721 | 117 | ||
| AY333960.1 | - | 9 - 10 | 678 | 117 | ||
| BC019146.1 | - | 9 - 10 | 707 | 5052822 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpld1
Genomic Map