Assay Details
| Gene Symbol: Pak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011035.2 | NP_035165.2 | 11 - 12 | 1341 | 75 | |
| GenBank mRNA | AF082077.1 | - | 11 - 12 | 1306 | 75 | |
| BC055740.1 | - | 8 - 9 | 989 | 75 | ||
| BC060157.1 | - | 8 - 9 | 989 | 75 | ||
| BC079838.1 | - | 11 - 12 | 1326 | 75 | ||
| BC145698.1 | - | 12 - 13 | 1265 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak1
Genomic Map