Assay Details
| Gene Symbol: Pak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011035.2 | NP_035165.2 | 12 - 13 | 1443 | 68 | |
| GenBank mRNA | AF082077.1 | - | 12 - 13 | 1408 | 68 | |
| AK077497.1 | - | 12 - 13 | 1408 | 68 | ||
| BC055740.1 | - | 9 - 10 | 1091 | 68 | ||
| BC060157.1 | - | 9 - 10 | 1091 | 68 | ||
| BC079838.1 | - | 12 - 13 | 1428 | 68 | ||
| BC145698.1 | - | 13 - 14 | 1367 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak1
Genomic Map