Assay Details
| Gene Symbol: Pak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011035.2 | NP_035165.2 | 13 - 14 | 1633 | 99 | |
| GenBank mRNA | AF082077.1 | - | 13 - 14 | 1598 | 99 | |
| AK077497.1 | - | 13 - 14 | 1598 | 99 | ||
| BC055740.1 | - | 10 - 11 | 1281 | 99 | ||
| BC060157.1 | - | 10 - 11 | 1281 | 99 | ||
| BC079838.1 | - | 13 - 14 | 1618 | 99 | ||
| BC145698.1 | - | 14 - 15 | 1557 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak1
Genomic Map