Assay Details
| Gene Symbol: Pak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011035.2 | NP_035165.2 | 2 - 3 | 411 | 105 | |
| GenBank mRNA | AK077497.1 | - | 2 - 3 | 377 | 105 | |
| BC055740.1 | - | 2 - 3 | 415 | 105 | ||
| BC060157.1 | - | 2 - 3 | 415 | 105 | ||
| BC079838.1 | - | 2 - 3 | 397 | 105 | ||
| BC145698.1 | - | 3 - 4 | 335 | 105 | ||
More Information
Set Membership: |
|

Pak1
Genomic Map