Assay Details
| Gene Symbol: Pak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011035.2 | NP_035165.2 | 4 - 5 | 663 | 134 | |
| GenBank mRNA | AF082077.1 | - | 4 - 5 | 625 | 134 | |
| AK077497.1 | - | 4 - 5 | 629 | 134 | ||
| BC079838.1 | - | 4 - 5 | 649 | 134 | ||
| BC145698.1 | - | 5 - 6 | 587 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak1
Genomic Map