Assay Details
| Gene Symbol: Pak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011035.2 | NP_035165.2 | 5 - 6 | 706 | 126 | |
| GenBank mRNA | AK077497.1 | - | 5 - 6 | 672 | 126 | |
| BC079838.1 | - | 5 - 6 | 692 | 126 | ||
| BC145698.1 | - | 6 - 7 | 630 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak1
Genomic Map