Assay Details
| Gene Symbol: Pak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011035.2 | NP_035165.2 | 8 - 9 | 1061 | 70 | |
| GenBank mRNA | AK077497.1 | - | 8 - 9 | 1027 | 70 | |
| BC145698.1 | - | 9 - 10 | 985 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak1
Genomic Map