Assay Details
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027629.3 | NP_081905.1 | 6 - 7 | 1067 | 126 | |
GenBank mRNA | AK016437.1 | - | 4 - 5 | 426 | 126 | |
AK039189.1 | - | 6 - 7 | 1051 | 126 | ||
BC020351.1 | - | 6 - 7 | 1280 | 126 | ||
BC032150.1 | - | 2 - 3 | 273 | 5004308 | 126 | |
BC076571.1 | - | 6 - 7 | 964 | 30356163 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
