Assay Details
| Gene Symbol: Trim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285870.1 | NP_001272799.1 | 3 - 4 | 593 | 60 | |
| NM_001285871.1 | NP_001272800.1 | 3 - 4 | 778 | 60 | ||
| NM_001285873.1 | NP_001272802.1 | 3 - 4 | 593 | 60 | ||
| NM_018880.3 | NP_061368.1 | 3 - 4 | 567 | 60 | ||
| GenBank mRNA | AB030912.1 | - | 3 - 4 | 594 | 60 | |
| AF220019.1 | - | 3 - 4 | 486 | 60 | ||
| AK019165.1 | - | 3 - 4 | 528 | 60 | ||
| AK144674.1 | - | 3 - 4 | 496 | 60 | ||
| AK170029.1 | - | 3 - 4 | 531 | 60 | ||
| AK196980.1 | - | 1 - 2 | 177 | 60 | ||
| BC003347.1 | - | 3 - 4 | 608 | 60 | ||
| BC034263.1 | - | 3 - 4 | 532 | 4503884 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim3
Genomic Map