Assay Details
| Gene Symbol: Tcaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014379.1 | - | 1 - 2 | 160 | 69 | |
| AK083917.1 | - | 1 - 2 | 78 | 69 | ||
| AK132148.1 | - | 1 - 2 | 98 | 69 | ||
| AK173013.1 | - | 1 - 2 | 112 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014379.1, AK083917.1, AK132148.1 transcripts.
Set Membership: |
|

Tcaf1
Genomic Map