Assay Details
| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028123.3 | NP_082399.3 | 9 - 10 | 1043 | 110 | |
| GenBank mRNA | AK076213.1 | - | 9 - 10 | 986 | 110 | |
| AK077836.1 | - | 9 - 10 | 987 | 110 | ||
| AK145242.1 | - | 9 - 10 | 1013 | 110 | ||
| AK167716.1 | - | 9 - 10 | 999 | 110 | ||
| BC005744.1 | - | 9 - 10 | 862 | 110 | ||
| BC099673.1 | - | 10 - 11 | 980 | 4936456 | 110 | |
| BC100391.1 | - | 10 - 11 | 1077 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc37a3
Genomic Map