Assay Details
| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028123.3 | NP_082399.3 | 11 - 12 | 1283 | 63 | |
| GenBank mRNA | AK012071.1 | - | 3 - 4 | 506 | 63 | |
| AK076213.1 | - | 11 - 12 | 1226 | 63 | ||
| AK077836.1 | - | 11 - 12 | 1226 | 63 | ||
| AK145242.1 | - | 11 - 12 | 1253 | 63 | ||
| BC005744.1 | - | 11 - 12 | 1102 | 63 | ||
| BC099673.1 | - | 12 - 13 | 1220 | 4936456 | 63 | |
| BC100391.1 | - | 12 - 13 | 1317 | 63 | ||
More Information
Set Membership: |
|

Slc37a3
Genomic Map