Assay Details
| Gene Symbol: Ppfibp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008905.2 | NP_032931.2 | 1 - 2 | 330 | 75 | |
| GenBank mRNA | AK035364.1 | - | 1 - 2 | 98 | 75 | |
| AK171349.1 | - | 1 - 2 | 175 | 75 | ||
| BB627669.1 | - | 1 - 2 | 95 | 75 | ||
| BC051202.1 | - | 1 - 2 | 178 | 75 | ||
| CJ075536.1 | - | 1 - 2 | 336 | 75 | ||
| U79024.1 | - | 1 - 2 | 115 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK035364.1, NM_008905.2, U79024.1 transcripts.
Set Membership: |
|

Ppfibp2
Genomic Map