| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700013231.mm.1 |
TC0700001581.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700013231.mm.1 |
NM_001163557.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700013231.mm.1 |
NM_001163557.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700013231.mm.1 |
NM_008905.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700013231.mm.1 |
NM_008905.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700013230.mm.1 |
TC0700001581.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700013230.mm.1 |
NM_001163557.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700013230.mm.1 |
NM_001163557.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700013230.mm.1 |
NM_008905.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700013230.mm.1 |
NM_008905.2 |
Refseq |