Assay Details
| Gene Symbol: Trim24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272064.1 | NP_001258993.1 | 10 - 11 | 1905 | 95 | |
| NM_001272076.1 | NP_001259005.1 | 10 - 11 | 1587 | 95 | ||
| NM_145076.4 | NP_659542.3 | 10 - 11 | 2007 | 95 | ||
| GenBank mRNA | AB221637.1 | - | 10 - 11 | 1707 | 95 | |
| AK133144.1 | - | 10 - 11 | 1587 | 95 | ||
| AK166538.1 | - | 6 - 7 | 964 | 95 | ||
| BC025482.1 | - | 1 - 2 | 102 | 95 | ||
| BC056959.1 | - | 10 - 11 | 1985 | 5698079 | 95 | |
| S78219.1 | - | 10 - 11 | 1905 | 95 | ||
| S78221.1 | - | 10 - 11 | 2007 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim24
Genomic Map