Assay Details
| Gene Symbol: Trim24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272064.1 | NP_001258993.1 | 7 - 8 | 1439 | 111 | |
| NM_001272076.1 | NP_001259005.1 | 7 - 8 | 1019 | 111 | ||
| NM_145076.4 | NP_659542.3 | 7 - 8 | 1439 | 111 | ||
| GenBank mRNA | AB221637.1 | - | 7 - 8 | 1139 | 111 | |
| AK133144.1 | - | 7 - 8 | 1019 | 111 | ||
| AK166538.1 | - | 3 - 4 | 396 | 111 | ||
| BC056959.1 | - | 7 - 8 | 1417 | 5698079 | 111 | |
| S78219.1 | - | 7 - 8 | 1439 | 111 | ||
| S78221.1 | - | 7 - 8 | 1439 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim24
Genomic Map