Assay Details
| Gene Symbol: Cpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025350.3 | NP_079626.2 | 4 - 5 | 727 | 62 | |
| GenBank mRNA | AK003088.1 | - | 4 - 5 | 512 | 62 | |
| BC052661.1 | - | 4 - 5 | 741 | 30050094 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpa1
Genomic Map