Assay Details
| Gene Symbol: Swap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009302.3 | NP_033328.3 | 6 - 7 | 1007 | 87 | |
| GenBank mRNA | AF053974.1 | - | 6 - 7 | 899 | 87 | |
| AK019882.1 | - | 6 - 7 | 951 | 87 | ||
| AK146773.1 | - | 6 - 7 | 999 | 87 | ||
| AK172990.1 | - | 6 - 7 | 944 | 87 | ||
| BC065136.1 | - | 6 - 7 | 949 | 30546814 | 87 | |
More Information
Set Membership: |
|

Swap70
Genomic Map