Assay Details
| Gene Symbol: Tspan12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173007.3 | NP_766595.1 | 4 - 5 | 490 | 120 | |
| GenBank mRNA | AK033554.1 | - | 3 - 4 | 325 | 120 | |
| AK050737.1 | - | 4 - 5 | 491 | 120 | ||
| BC065152.1 | - | 3 - 4 | 301 | 6808971 | 120 | |
| BC068240.1 | - | 3 - 4 | 352 | 3674585 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan12
Genomic Map