Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700014790.mm.1 |
TC0700001738.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014790.mm.1 |
NM_028955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014790.mm.1 |
NM_028955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014791.mm.1 |
TC0700001738.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014791.mm.1 |
NM_028955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014791.mm.1 |
NM_028955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014792.mm.1 |
TC0700001738.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014792.mm.1 |
NM_028955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014792.mm.1 |
NM_028955.1 |
Refseq |