Assay Details
| Gene Symbol: Sh2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081459.2 | NP_001074928.1 | 4 - 5 | 1726 | 120 | |
| NM_001289538.1 | NP_001276467.1 | 4 - 5 | 1726 | 120 | ||
| NM_001289539.1 | NP_001276468.1 | 4 - 5 | 1349 | 120 | ||
| NM_001289540.1 | NP_001276469.1 | 5 - 6 | 1486 | 120 | ||
| NM_001289541.1 | NP_001276470.1 | 4 - 5 | 1349 | 120 | ||
| NM_001289542.1 | NP_001276471.1 | 4 - 5 | 1726 | 120 | ||
| NM_011363.3 | NP_035493.2 | 4 - 5 | 1726 | 120 | ||
| NR_110346.1 | 3 - 4 | 1214 | 120 | |||
| GenBank mRNA | AA111114.1 | - | 2 - 3 | 134 | 120 | |
| AF020526.1 | - | 4 - 5 | 1420 | 120 | ||
| AF074329.1 | - | 3 - 4 | 1131 | 120 | ||
| AF380422.1 | - | 4 - 5 | 1420 | 120 | ||
| AF421138.1 | - | 4 - 5 | 1420 | 120 | ||
| AF421139.1 | - | 4 - 5 | 1420 | 120 | ||
| AK154655.1 | - | 4 - 5 | 1660 | 120 | ||
| AK168439.1 | - | 4 - 5 | 1321 | 120 | ||
| AK170444.1 | - | 4 - 5 | 1700 | 120 | ||
| AK171328.1 | - | 4 - 5 | 1734 | 120 | ||
| BC011422.1 | - | 3 - 4 | 1188 | 3156603 | 120 | |
| BC051978.1 | - | 4 - 5 | 1269 | 6401205 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh2b1
Genomic Map