Assay Details
| Gene Symbol: Ctbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170744.1 | NP_001164215.1 | 3 - 4 | 2030 | 72 | |
| NM_009980.4 | NP_034110.1 | 5 - 6 | 907 | 72 | ||
| GenBank mRNA | AB033123.1 | - | 5 - 6 | 772 | 72 | |
| AF016508.1 | - | 3 - 4 | 379 | 72 | ||
| AK134399.1 | - | 4 - 5 | 431 | 72 | ||
| AK146713.1 | - | 5 - 6 | 792 | 72 | ||
| AK147871.1 | - | 5 - 6 | 790 | 72 | ||
| BC013333.1 | - | 3 - 4 | 414 | 4019572 | 72 | |
| BC017520.1 | - | 4 - 5 | 1875 | 4511826 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctbp2
Genomic Map