Assay Details
| Gene Symbol: Tubgcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286007.1 | NP_001272936.1 | 10 - 11 | 1906 | 107 | |
| NM_001286009.1 | NP_001272938.1 | 10 - 11 | 1906 | 107 | ||
| NM_001286011.1 | NP_001272940.1 | 9 - 10 | 1673 | 107 | ||
| NM_133755.2 | NP_598516.2 | 10 - 11 | 1833 | 107 | ||
| GenBank mRNA | AK033157.1 | - | 10 - 11 | 1623 | 107 | |
| AK045084.1 | - | 10 - 11 | 1622 | 107 | ||
| AK146771.1 | - | 10 - 11 | 1828 | 107 | ||
| AK153511.1 | - | 10 - 11 | 1754 | 107 | ||
| AK158060.1 | - | 10 - 11 | 1622 | 107 | ||
| AK179002.1 | - | 2 - 3 | 292 | 107 | ||
| AK194815.1 | - | 1 - 2 | 99 | 107 | ||
| BC012519.1 | - | 10 - 11 | 1761 | 3601589 | 107 | |
| BC025564.1 | - | 9 - 10 | 1595 | 107 | ||
| BC025582.1 | - | 10 - 11 | 1722 | 5324981 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tubgcp2
Genomic Map