Assay Details
| Gene Symbol: Tubgcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286007.1 | NP_001272936.1 | 14 - 15 | 2513 | 61 | |
| NM_001286009.1 | NP_001272938.1 | 14 - 15 | 2513 | 61 | ||
| NM_001286011.1 | NP_001272940.1 | 13 - 14 | 2280 | 61 | ||
| NM_133755.2 | NP_598516.2 | 14 - 15 | 2440 | 61 | ||
| GenBank mRNA | AK045084.1 | - | 14 - 15 | 2229 | 61 | |
| AK146771.1 | - | 14 - 15 | 2435 | 61 | ||
| AK158060.1 | - | 14 - 15 | 2229 | 61 | ||
| AK167924.1 | - | 14 - 15 | 2335 | 61 | ||
| BC012519.1 | - | 14 - 15 | 2368 | 3601589 | 61 | |
| BC025564.1 | - | 13 - 14 | 2202 | 61 | ||
| BC025582.1 | - | 14 - 15 | 2329 | 5324981 | 61 | |
| BC027034.1 | - | 13 - 14 | 2714 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tubgcp2
Genomic Map