Assay Details
| Gene Symbol: Kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026370.1 | NP_080646.1 | 2 - 3 | 304 | 77 | |
| GenBank mRNA | AK003264.1 | - | 2 - 3 | 304 | 77 | |
| AK017161.1 | - | 1 - 2 | 50 | 77 | ||
| AK018002.1 | - | 2 - 3 | 301 | 77 | ||
| AK029350.1 | - | 2 - 3 | 306 | 77 | ||
| AK132000.1 | - | 2 - 3 | 305 | 77 | ||
| AK146710.1 | - | 2 - 3 | 306 | 77 | ||
| BC036284.1 | - | 2 - 3 | 312 | 5136304 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kat8
Genomic Map