Assay Details
| Gene Symbol: Tmem80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141950.1 | NP_001135422.1 | 4 - 5 | 396 | 70 | |
| NM_027797.3 | NP_082073.1 | 4 - 5 | 465 | 70 | ||
| GenBank mRNA | AK017460.1 | - | 4 - 5 | 466 | 70 | |
| AK134821.1 | - | 4 - 5 | 397 | 70 | ||
| AK169083.1 | - | 4 - 5 | 355 | 70 | ||
| BC022756.1 | - | 4 - 5 | 340 | 4922223 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem80
Genomic Map