Assay Details
| Gene Symbol: Tmem80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141950.1 | NP_001135422.1 | 5 - 6 | 489 | 76 | |
| NM_027797.3 | NP_082073.1 | 5 - 6 | 558 | 76 | ||
| GenBank mRNA | AK017460.1 | - | 5 - 6 | 559 | 76 | |
| AK134821.1 | - | 5 - 6 | 490 | 76 | ||
| AK169083.1 | - | 5 - 6 | 448 | 76 | ||
| BC022756.1 | - | 5 - 6 | 433 | 4922223 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588180 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588179 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960298 | AK134821.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960298 | AK169083.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960298 | NM_027797.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960298 | AK052566.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960298 | NM_001141950.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178632 | AK148367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178632 | NR_024509.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117641 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960298 | AK134821.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960298 | AK169083.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960298 | NM_027797.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960298 | AK052566.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960298 | NM_001141950.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178632 | AK148367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178632 | NR_024509.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25588191 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117641 | - | - |
| Whole Mouse Genome G4122A | A_52_P105249 | AK134821.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P105249 | AK169083.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P105249 | BC022756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P105249 | NM_027797.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P105249 | AK017460.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119730 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117641 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05900945 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05900932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588180 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25588179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25588277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06217004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06217012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05900947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06183971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06441961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06026642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06256475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06216995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05900945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06183975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05900938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06216998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06221446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06183970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25588223 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10558878 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558874 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558879 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558877 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558875 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431726_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10558872 | - | - |
| Mouse Genome 430 2.0 Array | 1431726_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1431726_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71448_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71448_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700018143.mm.1 | TC0700002046.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700018143.mm.1 | NM_001141950.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018143.mm.1 | NM_001141950.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018143.mm.1 | NM_027797.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018143.mm.1 | NM_027797.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018149.mm.1 | TC0700002046.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700018149.mm.1 | NM_001141950.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018149.mm.1 | NM_001141950.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018149.mm.1 | NM_027797.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018149.mm.1 | NM_027797.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018145.mm.1 | TC0700002046.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700018145.mm.1 | NM_001141950.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018145.mm.1 | NM_001141950.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018145.mm.1 | NM_027797.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018145.mm.1 | NM_027797.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018146.mm.1 | TC0700002046.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700018146.mm.1 | NM_001141950.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018146.mm.1 | NM_001141950.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018146.mm.1 | NM_027797.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018146.mm.1 | NM_027797.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018144.mm.1 | TC0700002046.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700018144.mm.1 | NM_001141950.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018144.mm.1 | NM_001141950.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018144.mm.1 | NM_027797.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018144.mm.1 | NM_027797.3 | Refseq |
| Mouse Expression Set 430 | 1431726_a_at | - | - |
| Mouse Expression Set 430 | 1431726_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431726_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431726_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431726_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431726_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1990059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1990059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102450301 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001990059 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218554 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2456669 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218554 | - | - |
| mouse-6 v1.1 (Illumina) | scl31888.5_427-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0071448.2_44-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102450301 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1990059 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0071448.2_44-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102450301 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990059 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2622780 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2622780 | - | - |

Tmem80
Genomic Map