Assay Details
| Gene Symbol: Cep41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ278891.1 | - | 8 - 9 | 688 | 92 | |
| AK132495.1 | - | 8 - 9 | 677 | 92 | ||
| AK161461.1 | - | 8 - 9 | 662 | 92 | ||
| AK166974.1 | - | 9 - 10 | 805 | 92 | ||
| AK170213.1 | - | 8 - 9 | 701 | 92 | ||
| BC031892.1 | - | 8 - 9 | 658 | 5365552 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK166974.1 transcripts.
Set Membership: |
|

Cep41
Genomic Map