Assay Details
| Gene Symbol: Tspan33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301407.1 | NP_001288336.1 | 3 - 4 | 512 | 88 | |
| NM_146173.3 | NP_666285.1 | 3 - 4 | 515 | 88 | ||
| GenBank mRNA | AF276890.1 | - | 3 - 4 | 514 | 88 | |
| AK004954.1 | - | 3 - 4 | 489 | 88 | ||
| AK162631.1 | - | 2 - 3 | 2110 | 88 | ||
| AK172176.1 | - | 3 - 4 | 493 | 88 | ||
| BC024685.1 | - | 3 - 4 | 473 | 3981492 | 88 | |
| BE861472.1 | - | 1 - 2 | 124 | 88 | ||
| BU054333.1 | - | 3 - 4 | 266 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tspan33
Genomic Map