Assay Details
| Gene Symbol: Tspan33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301407.1 | NP_001288336.1 | 5 - 6 | 686 | 99 | |
| NM_146173.3 | NP_666285.1 | 5 - 6 | 689 | 99 | ||
| GenBank mRNA | AF276890.1 | - | 5 - 6 | 688 | 99 | |
| AK172176.1 | - | 5 - 6 | 667 | 99 | ||
| BC024685.1 | - | 5 - 6 | 647 | 3981492 | 99 | |
| BE861472.1 | - | 3 - 4 | 298 | 99 | ||
| BU054333.1 | - | 5 - 6 | 440 | 99 | ||
More Information
Set Membership: |
|

Tspan33
Genomic Map