Assay Details
| Gene Symbol: Hirip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172746.3 | NP_766334.1 | 5 - 6 | 2021 | 59 | |
| GenBank mRNA | AK045122.1 | - | 5 - 6 | 1556 | 59 | |
| AK135526.1 | - | 5 - 6 | 1566 | 59 | ||
| AK161657.1 | - | 5 - 6 | 1556 | 59 | ||
| AK162096.1 | - | 2 - 3 | 583 | 59 | ||
| BC055687.1 | - | 5 - 6 | 2021 | 6414655 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hirip3
Genomic Map