Assay Details
| Gene Symbol: Srebf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011480.3 | NP_035610.1 | 9 - 10 | 1810 | 128 | |
| GenBank mRNA | AF374266.1 | - | 8 - 9 | 1574 | 128 | |
| AK052628.1 | - | 9 - 10 | 1811 | 128 | ||
| AK150052.1 | - | 9 - 10 | 1799 | 128 | ||
| AK154424.1 | - | 9 - 10 | 1778 | 128 | ||
| AK158269.1 | - | 2 - 3 | 359 | 128 | ||
| AK169607.1 | - | 9 - 10 | 1811 | 128 | ||
| BC006051.1 | - | 6 - 7 | 946 | 128 | ||
| BC056922.1 | - | 9 - 10 | 1792 | 6824948 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srebf1
Genomic Map