Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100026125.mm.1 |
NM_001039092.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026125.mm.1 |
NM_001039092.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026125.mm.1 |
NM_001039093.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026125.mm.1 |
NM_001039093.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026125.mm.1 |
NM_153080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026125.mm.1 |
NM_153080.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026125.mm.1 |
TC1100002871.mm.1 |
MTA TC |