Assay Details
| Gene Symbol: Pmpcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028431.2 | NP_082707.1 | 3 - 4 | 343 | 84 | |
| GenBank mRNA | AK013995.1 | - | 3 - 4 | 344 | 84 | |
| AK152673.1 | - | 3 - 4 | 344 | 84 | ||
| AK169417.1 | - | 3 - 4 | 300 | 84 | ||
| BC038321.1 | - | 2 - 3 | 214 | 84 | ||
| BC051216.1 | - | 2 - 3 | 150 | 84 | ||
| BC060662.1 | - | 3 - 4 | 336 | 84 | ||
| BC065095.1 | - | 3 - 4 | 336 | 84 | ||
| BC087943.1 | - | 3 - 4 | 355 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pmpcb
Genomic Map