Assay Details
| Gene Symbol: Pmpcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028431.2 | NP_082707.1 | 7 - 8 | 871 | 59 | |
| GenBank mRNA | AK013995.1 | - | 7 - 8 | 872 | 59 | |
| AK152673.1 | - | 7 - 8 | 872 | 59 | ||
| AK169417.1 | - | 7 - 8 | 828 | 59 | ||
| BC038321.1 | - | 6 - 7 | 742 | 59 | ||
| BC051216.1 | - | 6 - 7 | 678 | 59 | ||
| BC060662.1 | - | 6 - 7 | 3383 | 59 | ||
| BC065095.1 | - | 6 - 7 | 3383 | 59 | ||
| BC087943.1 | - | 7 - 8 | 883 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pmpcb
Genomic Map