Assay Details
| Gene Symbol: Folr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252552.1 | NP_001239481.1 | 3 - 4 | 482 | 67 | |
| NM_001252553.1 | NP_001239482.1 | 3 - 4 | 408 | 67 | ||
| NM_001252554.1 | NP_001239483.1 | 3 - 4 | 395 | 67 | ||
| NM_008034.3 | NP_032060.2 | 3 - 4 | 479 | 67 | ||
| GenBank mRNA | AF096319.1 | - | 3 - 4 | 374 | 67 | |
| AF096320.1 | - | 1 - 2 | 269 | 67 | ||
| BC037078.1 | - | 2 - 3 | 220 | 67 | ||
| BC138781.1 | - | 2 - 3 | 112 | 8861803 | 67 | |
| BC138795.1 | - | 2 - 3 | 199 | 8861817 | 67 | |
| BC145414.1 | - | 2 - 3 | 199 | 9053972 | 67 | |
| CD241494.1 | - | 3 - 4 | 492 | 67 | ||
| CF617679.1 | - | 2 - 3 | 225 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF096319.1, AF096320.1, BC037078.1, BC138781.1, BC138795.1, BC145414.1, NM_001252552.1, NM_001252553.1, NM_001252554.1, NM_008034.3 transcripts.
Set Membership: |
|

Folr1
Genomic Map