Assay Details
| Gene Symbol: Spaca6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162909.1 | NP_001156381.1 | 2 - 3 | 370 | 116 | |
| GenBank mRNA | AK016052.1 | - | 2 - 3 | 282 | 116 | |
| BC171982.1 | - | 2 - 3 | 165 | 9054268 | 116 | |
| BC171984.1 | - | 2 - 3 | 165 | 9054270 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC171982.1, BC171984.1 transcripts.
Set Membership: |
|

Spaca6
Genomic Map