Assay Details
| Gene Symbol: Spaca6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162909.1 | NP_001156381.1 | 5 - 6 | 540 | 66 | |
| GenBank mRNA | AK016052.1 | - | 5 - 6 | 452 | 66 | |
| BC171982.1 | - | 5 - 6 | 335 | 9054268 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC171982.1 transcripts.
Set Membership: |
|

Spaca6
Genomic Map