Assay Details
| Gene Symbol: Spaca6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162909.1 | NP_001156381.1 | 7 - 8 | 801 | 71 | |
| GenBank mRNA | AK016052.1 | - | 7 - 8 | 711 | 71 | |
| AK080293.1 | - | 2 - 3 | 238 | 71 | ||
| BC171977.1 | - | 7 - 8 | 565 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spaca6
Genomic Map